Learn More
Medchemexpress LLC HY-101428 5mg Medchemexpress, NS-638 CAS:150493-34-8 Purity:>98%
SDP
Catalog No. 501932125
Encompass_Preferred
Medchemexpress, HY-101428 5mg NS-638 CAS:150493-34-8 NS-638 is a small nonpeptide molecule with Ca 2+ -channel blocking properties. K + -stimulated intracellular Ca 2+ -elevation is blocked with an IC 50 value of 3.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2125 Supplier Medchemexpress LLC Supplier No. HY1014285MG
Medchemexpress, HY-101428 5mg NS-638 CAS:150493-34-8 NS-638 is a small nonpeptide molecule with Ca 2+ -channel blocking properties. K + -stimulated intracellular Ca 2+ -elevation is blocked with an IC 50 value of 3.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.