Learn More
Cayman Chemical NS 8593 (hydrochloride), Purity: ≥98%, CAS: 875755-24-1, 10 mg
SDP
Catalog No. 502990214
Encompass_Preferred
An inhibitory gating modifier of SK channels (Kds = 0.42, 0.6, and 0.73 μM for SK1, SK2, and SK3, respectively, in the presence of calcium); decreases the calcium sensitivity of SK channels; selective for SK channels over IK and BK potassium channels at 10 μM; induces relaxation of potassium- or acetylcholine chloride-precontracted isolated tracheal rings from wild-type mice (IC50s = 8.9 and 39.8 μM, respectively) or from mice in an ovalbumin-induced model of allergic asthma (IC50s = 16.4 and 32.2 μM, respectively); inhibits aerosolized acetylcholine chloride-induced increases in respiratory system resistance in mice when administered as an aerosol at 500 μM; decreases the duration of burst-pacing-induced atrial fibrillation in normotensive and spontaneously hypertensive rats at 5 mg/kg
Catalog No. 50-299-0214 Supplier Cayman Chemical Supplier No. 2977410 MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
An inhibitory gating modifier of SK channels (Kds = 0.42, 0.6, and 0.73 μM for SK1, SK2, and SK3, respectively, in the presence of calcium); decreases the calcium sensitivity of SK channels; selective for SK channels over IK and BK potassium channels at 10 μM; induces relaxation of potassium- or acetylcholine chloride-precontracted isolated tracheal rings from wild-type mice (IC50s = 8.9 and 39.8 μM, respectively) or from mice in an ovalbumin-induced model of allergic asthma (IC50s = 16.4 and 32.2 μM, respectively); inhibits aerosolized acetylcholine chloride-induced increases in respiratory system resistance in mice when administered as an aerosol at 500 μM; decreases the duration of burst-pacing-induced atrial fibrillation in normotensive and spontaneously hypertensive rats at 5 mg/kg