Learn More
TWD Scientific LLC Blunt Tip Disposable Forceps, White. Pack of 100
SDP
Catalog No. NC9978916 Shop All TWD Scientific LLC Products
Encompass_Preferred
Serrated blunt tip disposable forceps, reinforced nylon, non sterile, autoclavable, white. Length: 4.9". 100/Pack
Catalog No. NC9978916 Supplier TWD Scientific LLC Supplier No. DF8088N
Serrated blunt tip disposable forceps, reinforced nylon, non sterile, autoclavable, white. Length: 4.9". 100/Pack