Learn More
ACROBiosystems ACROBiosystems Zika virus NS1 Protein, His Tag
Supplier:  ACROBiosystems NS1Z522250UG
Encompass_Preferred
Recombinant Protein;50UG;Zika virus NS1, His Tag (NS1-Z5222) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.0 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7311