Learn More
Alkali Scientific Alkali Scientific™ NS1000 SmartDrop L, Nano Spectrophotometer For Basic DNA/RNA/Protein Quantification And Purity, 115V
SDP
Catalog No. 503029660
Encompass_Preferred
Ideal for basic DNA/RNA/protein quantification and purity Affordable LED and silicon detector technology Fixed 0.5mm path length, detect up to 2500ng/µl of dsDNA SmartDrop L, NS1000 is the affordable choice for laboratories needing to determine nucleic acid and protein concentrations and purity of samples. The LED light source and silicon photocell sensor provide readings at 260 and 280nm. Samples from 1.0 to 2.0µl are loaded directly on the pedestal. Results are provided in about 5 seconds. The software automaticallycalculates and displays 260/280 ratios.
Catalog No. 50-302-9660 Supplier Alkali Scientific Supplier No. SD1000
Add to Cart
Add to Cart
Request a Quote
Ideal for basic DNA/RNA/protein quantification and purity Affordable LED and silicon detector technology Fixed 0.5mm path length, detect up to 2500ng/µl of dsDNA SmartDrop L, NS1000 is the affordable choice for laboratories needing to determine nucleic acid and protein concentrations and purity of samples. The LED light source and silicon photocell sensor provide readings at 260 and 280nm. Samples from 1.0 to 2.0µl are loaded directly on the pedestal. Results are provided in about 5 seconds. The software automaticallycalculates and displays 260/280 ratios.