Learn More
Alkali Scientific Alkali Scientific™ NS1010 SmartDrop X, Nano Spectrophotometer With Full Spectrum Absorbance Scans And Auto-Ranging Path Length, 115V
SDP
Catalog No. 503029503
Encompass_Preferred
Full-spectrum absorbance scans from 200 to 800nm Quickly calculates and displays 260/280 and 260/230 ratios Long-life Xenon lamp with precision monochromator Auto-ranging path lengths of 0.2mm and 1.0mm Detect up to 4,500 ng/µl of dsDNA without dilution A full spectrum UV-Vis spectrophotometer, SmartDrop X NS1010 requires 0.5 to 2.0µl of sample. In addition to nucleic acid and protein calculations, the software has preset programs for BCA, Bradford and Lowry assays. A full spectrum analysis can be performed in about 6 seconds, aiding in the identification of unknowns. Up to 5 specific points on the spectrum can be selected for colorimetric readings output. The long life Xenon lamp source requires no warm up and is paried with a highly sensitive CCD detector. To provide the most accurate results, the SmartDrop X utilizes 2 path lengths.
Catalog No. 50-302-9503 Supplier Alkali Scientific Supplier No. SD1010
Add to Cart
Add to Cart
Request a Quote
Full-spectrum absorbance scans from 200 to 800nm Quickly calculates and displays 260/280 and 260/230 ratios Long-life Xenon lamp with precision monochromator Auto-ranging path lengths of 0.2mm and 1.0mm Detect up to 4,500 ng/µl of dsDNA without dilution A full spectrum UV-Vis spectrophotometer, SmartDrop X NS1010 requires 0.5 to 2.0µl of sample. In addition to nucleic acid and protein calculations, the software has preset programs for BCA, Bradford and Lowry assays. A full spectrum analysis can be performed in about 6 seconds, aiding in the identification of unknowns. Up to 5 specific points on the spectrum can be selected for colorimetric readings output. The long life Xenon lamp source requires no warm up and is paried with a highly sensitive CCD detector. To provide the most accurate results, the SmartDrop X utilizes 2 path lengths.