Learn More
Alkali Scientific NS1020 SmartDrop XF Nano Full
SDP
Catalog No. 503029502
Encompass_Preferred
Full-spectrum absorbance scanning and calculations of260/280 and 260/230 ratios Long-life Xenon lamp with precision monochromator Auto-ranging path lengths of 0 05mm 0 2mm and1 0mm Detect up to 15000ng/ l of dsDNA without dilution Integrated fluorometer for sensitive quantitation of lowconcentration nucleic acids This top of the line spectrophotometer NS1020 can perform not only UV-Vis readings in the range of 200-800nm but also fluorescence readings Like the SmartDrop X the SmartDrop XF features predefined programs for nucleic acids proteins and various colorimetric assays The patented motorized system gently stretches the sample column and allows for three different path lengths providing accurate readings of samples from 0 5 to 2 0 l without dilution The integrated fluorometer yields more accurate sensitive and selective quantitation of nucleic acids The blue light channel (460nm) is useful for detection of dyes such as PicoGreen and RiboGreen as well as GFP and fluorescein
Catalog No. 50-302-9502 Supplier Alkali Scientific Supplier No. SD1020
Add to Cart
Add to Cart
Request a Quote
Full-spectrum absorbance scanning and calculations of260/280 and 260/230 ratios Long-life Xenon lamp with precision monochromator Auto-ranging path lengths of 0 05mm 0 2mm and1 0mm Detect up to 15000ng/ l of dsDNA without dilution Integrated fluorometer for sensitive quantitation of lowconcentration nucleic acids This top of the line spectrophotometer NS1020 can perform not only UV-Vis readings in the range of 200-800nm but also fluorescence readings Like the SmartDrop X the SmartDrop XF features predefined programs for nucleic acids proteins and various colorimetric assays The patented motorized system gently stretches the sample column and allows for three different path lengths providing accurate readings of samples from 0 5 to 2 0 l without dilution The integrated fluorometer yields more accurate sensitive and selective quantitation of nucleic acids The blue light channel (460nm) is useful for detection of dyes such as PicoGreen and RiboGreen as well as GFP and fluorescein