Learn More
Medchemexpress LLC HY-102070 5mg Medchemexpress, NS13001 CAS:1063331-94-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020705MG
Encompass_Preferred
Medchemexpress, HY-102070 5mg NS13001 CAS:1063331-94-1 NS13001 is a potent, selective, orally active allosteric positive modulator of SK channels (small conductance calcium-activated potassium channels). The EC50s are 1.8 and 0.14 μM for SK2 and SK3, respectively. NS13001 holds promise as a potential therapeutic agent for treatment of spinocerebellar ataxia type 2 (SCA2) and possibly other cerebellar ataxias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7978