Learn More
TARGETMOL CHEMICALS INC NS1643 50MG
SDP
Catalog No. 502580344
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NS1643 is a potent human ether-a-go-go related gene (hERG) KV11.1 channel activator with EC50 of 10.5 uM. purity: 99%
Catalog No. 50-258-0344 Supplier TARGETMOL CHEMICALS INC Supplier No. T319250MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NS1643 is a potent human ether-a-go-go related gene (hERG) KV11.1 channel activator with EC50 of 10.5 uM. purity: 99%