Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NS2B/NS3-IN-3 hydrochloride | 2832876-91-0 | 97.0% | C19H22CIN3O2 | 1 ML
SDP

Catalog No. 5000354785
Encompass_Preferred

NS2B/NS3-IN-3 hydrochloride is an inhibitor of Flavivirus NS2B-NS3 protease, exhibiting strong antiviral activity against cellular replication of Zika virus in cells with EC68 values of 1-3 μM. This chemical appears as an orange to red solid and has a purity of 97.0%.

  • Inhibits flavivirus NS2B-NS3 protease
  • Strong antiviral activity against cellular replication of Zika virus
  • Molecular weight: 359.85
  • Storage at 4°C, protected from light and under nitrogen
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Catalog No. 50-003-54785 Supplier Medchemexpress LLC Supplier No. HY144644A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NS2B/NS3-IN-3 hydrochloride is an inhibitor of Flavivirus NS2B-NS3 protease, exhibiting strong antiviral activity against cellular replication of Zika virus in cells with EC68 values of 1-3 μM. This chemical appears as an orange to red solid and has a purity of 97.0%.

  • Inhibits flavivirus NS2B-NS3 protease
  • Strong antiviral activity against cellular replication of Zika virus
  • Molecular weight: 359.85
  • Storage at 4°C, protected from light and under nitrogen
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.