Learn More
Abcam NS309, K+ channel activator, 50MG
Catalog No. 502707641
Encompass_Preferred
MW 231.03 Da, Purity >98%. Potent activator of human Ca2+ -activated K+ channels of SK and IK types, selective over BK type channels (EC50 = 10 nM for increasing hIK current). At least 1000 times more potent than 1-EBIO (ab120258). Can be used in conjunction with CyPPA (ab120829) to distinguish SK1 and SK3 subtypes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7641 Supplier Abcam Supplier No. AB12037150MG
MW 231.03 Da, Purity >98%. Potent activator of human Ca2+ -activated K+ channels of SK and IK types, selective over BK type channels (EC50 = 10 nM for increasing hIK current). At least 1000 times more potent than 1-EBIO (ab120258). Can be used in conjunction with CyPPA (ab120829) to distinguish SK1 and SK3 subtypes.
The product is subject to the following: Abcam Restricted Use Statement