Learn More
Medchemexpress LLC NS3861 fumarate | 216853-59-7 | 99.8% | 284.22 | 1 ML
SDP
Catalog No. 5000343490
Encompass_Preferred
NS3861 is an agonist of nicotinic acetylcholine receptors (nAChRs) that binds with high affinity to the heteromeric α3β4 nAChR. It is intended for research use only.
  • Selectively activates α3-containing nAChRs.
  • Displays higher efficacy at the α3β2 receptor compared to the α3β4 receptor.
  • Exhibits high affinity and partial agonist properties in α3β4-expressed nAChRs.
  • Appears as an off-white to light yellow solid.
  • Soluble in DMSO at 50 mg/mL.
Catalog No. 50-003-43490 Supplier Medchemexpress LLC Supplier No. HY11012110MM/1ML
NS3861 is an agonist of nicotinic acetylcholine receptors (nAChRs) that binds with high affinity to the heteromeric α3β4 nAChR. It is intended for research use only.
  • Selectively activates α3-containing nAChRs.
  • Displays higher efficacy at the α3β2 receptor compared to the α3β4 receptor.
  • Exhibits high affinity and partial agonist properties in α3β4-expressed nAChRs.
  • Appears as an off-white to light yellow solid.
  • Soluble in DMSO at 50 mg/mL.