Learn More
Medchemexpress LLC HY-108588 10mg Medchemexpress, NS5806 CAS:426834-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10858810MG
Encompass_Preferred
Medchemexpress, HY-108588 10mg NS5806 CAS:426834-69-7 NS5806, a potent potassium current activator, increases KV4.3/KChIP2 peak current amplitudes with an EC50 of 5.3 μM. NS5806 slows KV4.3 and KV4.2 current dacay in channel complexes containing KChIP2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8584