Learn More
AdipoGen anti-NS5B (HCV), mAb (blocking) (5B-12B7) (100 µg)
SDP
Supplier:  AdipoGen AG20B0003C100
Encompass_Preferred
Monoclonal Antibody. Recognizes HCV NS5B from HCV genotype 1 (epitope mapped to NS5B aa 139-392). Does not react with genotype 2. Isotype: Mouse IgG2a. Clone: 5B-12B7. Applications: FUNC (Blocking), ICC, IP. Liquid. In PBS containing 0.02% sodium azide. The NS3 serine protease, in association with NS4A, is responsible for the cleavages of the Hepatitis C virus protease complexes NS3-NS4A, NS4A-NS4B, NS4B-NS5A and NS5A-NS5B. NS5B is a RNA-dependent RNA polymerase that plays an essential role in the virus replication. Hepatitis C virus protease NS3-NS4A colocalizes with IPS-1/VISA/MAVS/Cardif in the mitochondrial membrane and has been shown to cleave the TLR3 adapter TRIF and Cardif to eliminate the antiviral signaling pathways. NS3-NS4A represents an important target for anti-HCV drugs.
Catalog No. 50-205-2900