Learn More
Medchemexpress LLC HY-110105 5mg Medchemexpress, NS8593 (hydrochloride) CAS:875755-24-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1101055MG
Encompass_Preferred
Medchemexpress, HY-110105 5mg NS8593 (hydrochloride) CAS:875755-24-1 NS8593 hydrochloride is a potent and selective small conductance Ca2+-activated K+ channels (SK channels) inhibitor. NS8593 hydrochloride reversibly inhibits SK3-mediated currents with a Kd value of 77 nM. NS8593 hydrochloride inhibits all the SK1-3 subtypes Ca2+-dependently (Kds of 0.42, 0.60, and 0.73 μM, respectively, at 0.5 μM Ca2+), and does not affect the Ca2+-activated K+ channels of intermediate and large conductance (hIK and hBK channels, respectively)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9674