Learn More
Medchemexpress LLC NS8593 hydrochloride | 875755-24-1 | 99.76% | 299.80 | 1 MG
SDP
Catalog No. 5000344968
Encompass_Preferred
NS8593 hydrochloride is a potent and selective inhibitor of small conductance Ca2+-activated K+ channels (SK channels). It reversibly inhibits SK3-mediated currents with a Kd value of 77 nM. NS8593 hydrochloride inhibits all SK1-3 subtypes in a Ca2+-dependent manner (Kds of 0.42, 0.60, and 0.73 μM, respectively, at 0.5 μM Ca2+), but does not affect intermediate and large conductance Ca2+-activated K+ channels (hIK and hBK channels, respectively). It is for research use only and not sold to patients.
  • Potent and selective small conductance Ca2+-activated K+ channels inhibitor
  • Reversibly inhibits SK3-mediated currents with a Kd value of 77 nM
  • Inhibits SK1-3 subtypes Ca2+-dependently
  • Does not affect intermediate and large conductance Ca2+-activated K+ channels
  • Affects firing rate and firing pattern of dopaminergic neurons in vivo in C57Bl/6 mice at doses of 3 and 10 mg/kg intravenously
Catalog No. 50-003-44968 Supplier Medchemexpress LLC Supplier No. HY11010510MM/1ML
NS8593 hydrochloride is a potent and selective inhibitor of small conductance Ca2+-activated K+ channels (SK channels). It reversibly inhibits SK3-mediated currents with a Kd value of 77 nM. NS8593 hydrochloride inhibits all SK1-3 subtypes in a Ca2+-dependent manner (Kds of 0.42, 0.60, and 0.73 μM, respectively, at 0.5 μM Ca2+), but does not affect intermediate and large conductance Ca2+-activated K+ channels (hIK and hBK channels, respectively). It is for research use only and not sold to patients.
  • Potent and selective small conductance Ca2+-activated K+ channels inhibitor
  • Reversibly inhibits SK3-mediated currents with a Kd value of 77 nM
  • Inhibits SK1-3 subtypes Ca2+-dependently
  • Does not affect intermediate and large conductance Ca2+-activated K+ channels
  • Affects firing rate and firing pattern of dopaminergic neurons in vivo in C57Bl/6 mice at doses of 3 and 10 mg/kg intravenously