Learn More
MSC NSA Arc Flash Hood, 40 cal/cm2, CAT 4, 1 ea
Catalog No. 5000793013
Encompass
The ArcGuard RevoLite Lift Front Hood provides 40 cal/cm² arc flash protection, designed to reduce bulk and weight for improved wearer comfort and reduced heat stress. It features indirect vents for enhanced ventilation and communication.
  • PureView faceshield offers greater range of visibility with anti-fog and scratch-resistant coating
  • Streamlined design reduces bulk and weight for improved comfort
  • Specially designed lens optimizes field of view without distortion and reduces interior reflections
  • Clear chin cup expands the downward view
  • Lift front shield makes it easy to obtain fresh air while out of the hazard zone
  • Vents provide fresh air and increased auditory capabilities
  • FR fabric will not melt, drip, or ignite
  • Knit gussets at side provide greater range of motion and flexibility
  • Hard hat included
Catalog No. 50-007-93013 Supplier MSC Supplier No. 89948251
The ArcGuard RevoLite Lift Front Hood provides 40 cal/cm² arc flash protection, designed to reduce bulk and weight for improved wearer comfort and reduced heat stress. It features indirect vents for enhanced ventilation and communication.
  • PureView faceshield offers greater range of visibility with anti-fog and scratch-resistant coating
  • Streamlined design reduces bulk and weight for improved comfort
  • Specially designed lens optimizes field of view without distortion and reduces interior reflections
  • Clear chin cup expands the downward view
  • Lift front shield makes it easy to obtain fresh air while out of the hazard zone
  • Vents provide fresh air and increased auditory capabilities
  • FR fabric will not melt, drip, or ignite
  • Knit gussets at side provide greater range of motion and flexibility
  • Hard hat included