Learn More
Electron Microscopy Sciences NSA (Nonenyl Succinic Anhydride) 450 ML
SDP
Catalog No. 5027510
Encompass_Preferred
F.W. 227
Boiling point: 325 °C (617 °F)
An epoxy hardener that is specially distilled. The special distillation process offers you much clearer blocks than the standard NSA
Catalog No. 50-275-10 Supplier Electron Microscopy Sciences Supplier No. 19051
F.W. 227
Boiling point: 325 °C (617 °F)
An epoxy hardener that is specially distilled. The special distillation process offers you much clearer blocks than the standard NSA