Learn More
Cayman Chemical Ansamitocin P-3, Purity: ≥85%, CAS: 66547-09-9, 10 mg
SDP
Catalog No. 502983625
Encompass_Preferred
A microtubule depolymerizing agent that binds tubulin (Kd = 1.3 µM), depolymerizes microtubules in both interphase and mitosis, and perturbs chromosome segregation
Catalog No. 50-298-3625 Supplier Cayman Chemical Supplier No. 2053810 MG
A microtubule depolymerizing agent that binds tubulin (Kd = 1.3 µM), depolymerizes microtubules in both interphase and mitosis, and perturbs chromosome segregation