Learn More
Medchemexpress LLC NSC 107512 | 22242-89-3 | MFCD32671351 | 100.0% | C12H16N6O5 | 10MG
SDP
Catalog No. 5000260750
Encompass_Preferred
NSC 107512 is a sangivamycin-like small molecule inhibitor of cyclin-dependent kinase 9 (CDK9). It exhibits anti-proliferative activity and induces apoptosis in multiple myeloma models. This compound is supplied for laboratory research use only.
  • Potent CDK9 inhibitor with a sangivamycin-like scaffold.
  • Demonstrated to inhibit cell growth and induce apoptosis in multiple myeloma models.
  • High supplied purity (99.97%).
  • Molecular formula C12H16N6O5; molecular weight 324.29 g/mol.
  • Available as small-mass solid packs and as DMSO solution formats for convenience.
  • Suitable for long-term storage at -20°C (solid) and -80°C (in solvent) to preserve stability.
Catalog No. 50-002-60750 Supplier Medchemexpress LLC Supplier No. HY14168710MG
NSC 107512 is a sangivamycin-like small molecule inhibitor of cyclin-dependent kinase 9 (CDK9). It exhibits anti-proliferative activity and induces apoptosis in multiple myeloma models. This compound is supplied for laboratory research use only.
  • Potent CDK9 inhibitor with a sangivamycin-like scaffold.
  • Demonstrated to inhibit cell growth and induce apoptosis in multiple myeloma models.
  • High supplied purity (99.97%).
  • Molecular formula C12H16N6O5; molecular weight 324.29 g/mol.
  • Available as small-mass solid packs and as DMSO solution formats for convenience.
  • Suitable for long-term storage at -20°C (solid) and -80°C (in solvent) to preserve stability.