Learn More
Medchemexpress LLC NSC 135130 | 24164-06-5 | 97.1% | 245.32 | 250 MG
SDP
Catalog No. 5000402965
Encompass_Preferred
NSC 135130 (compound 11a) is a BOC-protected ADC linker that can be linked to tubulin-targeting inhibitors. It can be used to synthesize drug conjugates.
  • BOC-protected ADC linker
  • Links to tubulin-targeting inhibitors
  • Used to synthesize drug conjugates
Catalog No. 50-004-02965 Supplier Medchemexpress LLC Supplier No. 250MGHY41049
NSC 135130 (compound 11a) is a BOC-protected ADC linker that can be linked to tubulin-targeting inhibitors. It can be used to synthesize drug conjugates.
  • BOC-protected ADC linker
  • Links to tubulin-targeting inhibitors
  • Used to synthesize drug conjugates