Learn More
Medchemexpress LLC Nsc 146109 (hydrochloride) | 59474-01-0 | 99.9% | 316.85 | 25 MG
SDP
Catalog No. 5000347204
Encompass_Preferred
NSC 146109 hydrochloride is a small-molecule p53 activator that targets MDMX and can be used for breast cancer research. It is a pseudourea derivative that promotes breast cancer cells to undergo apoptosis by activating p53 and inducing the expression of proapoptotic genes.
  • Small-molecule p53 activator
  • Targets MDMX
  • Can be used for breast cancer research
  • Promotes apoptosis in breast cancer cells
  • Induces expression of proapoptotic genes
Catalog No. 50-003-47204 Supplier Medchemexpress LLC Supplier No. 25MGHY108638
NSC 146109 hydrochloride is a small-molecule p53 activator that targets MDMX and can be used for breast cancer research. It is a pseudourea derivative that promotes breast cancer cells to undergo apoptosis by activating p53 and inducing the expression of proapoptotic genes.
  • Small-molecule p53 activator
  • Targets MDMX
  • Can be used for breast cancer research
  • Promotes apoptosis in breast cancer cells
  • Induces expression of proapoptotic genes