Learn More
Medchemexpress LLC NSC 146109 (hydrochloride) | 59474-01-0 | MFCD09878281 | 99.6% | 316.85 g/mol | C17H17ClN2S | 5 MG
SDP
Catalog No. 5000263619
Encompass_Preferred
NSC 146109 hydrochloride is a research-grade small molecule that activates p53-dependent transcription and induces apoptosis in relevant cell models. Supplied as the hydrochloride salt, it is used in biochemical and cell-based assays to study p53 pathway modulation, MDMX targeting, and tumor suppressor activation.
  • High purity solid, white to yellow, suitable for research use.
  • Hydrochloride salt provides improved solubility for in vitro studies.
  • Demonstrated p53 activation and pro-apoptotic activity in cellular assays.
  • Available in small milligram quantities and ready-made DMSO solutions.
  • Storage recommendations support refrigerated and frozen stability.
Catalog No. 50-002-63619 Supplier Medchemexpress LLC Supplier No. HY1086385MG
NSC 146109 hydrochloride is a research-grade small molecule that activates p53-dependent transcription and induces apoptosis in relevant cell models. Supplied as the hydrochloride salt, it is used in biochemical and cell-based assays to study p53 pathway modulation, MDMX targeting, and tumor suppressor activation.
  • High purity solid, white to yellow, suitable for research use.
  • Hydrochloride salt provides improved solubility for in vitro studies.
  • Demonstrated p53 activation and pro-apoptotic activity in cellular assays.
  • Available in small milligram quantities and ready-made DMSO solutions.
  • Storage recommendations support refrigerated and frozen stability.