Learn More
Medchemexpress LLC NSC 14699 | 2513-33-9 | 99.3% | 258.23 | 250 MG
SDP
Catalog No. 5000403182
Encompass_Preferred
NSC 14699 is identified as a small molecule, non-peptide inhibitor specifically targeting botulinum neurotoxin serotype E. This compound holds potential for various research applications, particularly in the development of therapeutic and cosmetic agents.
  • Small molecule
  • Non-peptide inhibitor
  • Targets botulinum neurotoxin serotype E
  • Promising for research of therapeutic agents
  • Promising for research of cosmetic agents
Catalog No. 50-004-03182 Supplier Medchemexpress LLC Supplier No. 250MGHY42649
NSC 14699 is identified as a small molecule, non-peptide inhibitor specifically targeting botulinum neurotoxin serotype E. This compound holds potential for various research applications, particularly in the development of therapeutic and cosmetic agents.
  • Small molecule
  • Non-peptide inhibitor
  • Targets botulinum neurotoxin serotype E
  • Promising for research of therapeutic agents
  • Promising for research of cosmetic agents