Learn More
Selleck Chemical LLC NSC 207895 5mg 58131-57-0 XI-006
SDP
Supplier:  Selleck Chemical LLC S26785MG
Encompass_Preferred
NSC 207895 (XI-006) suppresses MDMX with IC50 of 2.5 ?M, leading to enhanced p53 stabilization/activation and DNA damage, and also regulates MDM2, an E3 ligase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2740