Learn More
Medchemexpress LLC HY-101084 100mg Medchemexpress, NSC 228155 CAS:113104-25-9 Purity:>98%
SDP
Catalog No. 501932029
Encompass_Preferred
Medchemexpress, HY-101084 100mg NSC 228155 CAS:113104-25-9 NSC 228155 is an activator of EGFR , binds to the extracellular region of EGFR and enhance tyrosine phosphorylation of EGFR [1] . NSC 228155 is also a potent inhibitor of KIX-KID interaction, inhibits kinase-inducible domain (KID) from CREB and KID-interacting domain (KIX) from CBP , with an IC 50 of 0.36 μM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2029 Supplier Medchemexpress LLC Supplier No. HY101084100MG
Medchemexpress, HY-101084 100mg NSC 228155 CAS:113104-25-9 NSC 228155 is an activator of EGFR , binds to the extracellular region of EGFR and enhance tyrosine phosphorylation of EGFR [1] . NSC 228155 is also a potent inhibitor of KIX-KID interaction, inhibits kinase-inducible domain (KID) from CREB and KID-interacting domain (KIX) from CBP , with an IC 50 of 0.36 μM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.