Learn More
Cayman Chemical NSC 23005 (sodium salt), Purity: ≥98%, CAS: 1796596-46-7, 1 mg
SDP
Catalog No. 502964140
Encompass_Preferred
An inhibitor of p18 that promotes expansion of murine and human HSCs; promotes HSC expansion in a single-cell in vitro culture assay using isolated murine bone marrow cells (ED50 = 5.21 nM); increases HSC proliferation without affecting differentiation, inducing cytotoxicity, or affecting leukemia cell proliferation
Catalog No. 50-296-4140 Supplier Cayman Chemical Supplier No. 233271 MG
An inhibitor of p18 that promotes expansion of murine and human HSCs; promotes HSC expansion in a single-cell in vitro culture assay using isolated murine bone marrow cells (ED50 = 5.21 nM); increases HSC proliferation without affecting differentiation, inducing cytotoxicity, or affecting leukemia cell proliferation