Learn More
Apexbio Technology LLC NSC 23766, 100mg. Cas: 1177865-17-6 MFCD: MFCD08703096. Selective inhibitor of Rac1-GEF interaction.
SDP
Supplier:  Apexbio Technology LLC A1952100
Encompass_Preferred
NSC-23766 is a specific inhibitor of Rac with IC50 of 50M.NSC-23766 blocks the activation of Rac 1 through binding the GEFs including Trio and Tiam 1 [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4728