Learn More
Selleck Chemical LLC NSC 23766 10mg 1177865-17-6
SDP
Supplier:  Selleck Chemical LLC S803110MG
Encompass_Preferred
NSC 23766 is an inhibitor of Rac GTPase targeting Rac activation by guanine nucleotide exchange factors (GEFs) with IC50 of ~50 ?M in a cell-free assay; does not inhibit the closely related targets, Cdc42 or RhoA. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5584