Learn More
Cayman Chemical NSC 23766hydrochlorIde 1mg
SDP
Catalog No. 502952741
Encompass_Preferred
A cell-permeable, reversible inhibitor of Rac1 activation by the Rac1-specific guanine nucleotide exchange factors TrioN and Tiam 1 (IC50 = 50 μM); has no effect on the closely related GTPases, Cdc42, and RhoA
Catalog No. 50-295-2741 Supplier Cayman Chemical Supplier No. 131961 MG
Add to Cart
Add to Cart
A cell-permeable, reversible inhibitor of Rac1 activation by the Rac1-specific guanine nucleotide exchange factors TrioN and Tiam 1 (IC50 = 50 μM); has no effect on the closely related GTPases, Cdc42, and RhoA