Learn More
Cayman Chemical NSC 23766 (hydrochloride), Purity: ≥98%, CAS: 1177865-17-6, 25 mg
SDP
Catalog No. 502952648
Encompass_Preferred
A cell-permeable, reversible inhibitor of Rac1 activation by the Rac1-specific guanine nucleotide exchange factors TrioN and Tiam 1 (IC50 = 50 μM); has no effect on the closely related GTPases, Cdc42, and RhoA
Catalog No. 50-295-2648 Supplier Cayman Chemical Supplier No. 1319625 MG
A cell-permeable, reversible inhibitor of Rac1 activation by the Rac1-specific guanine nucleotide exchange factors TrioN and Tiam 1 (IC50 = 50 μM); has no effect on the closely related GTPases, Cdc42, and RhoA