Learn More
Abcam NSC 26480 (5,6-dihydro-5-azacytidine), Nucleoside analog, 50MG
Catalog No. 502705503
Encompass_Preferred
MW 282.68 g/mol, Purity >99%. Nucleoside analog. 5-Azacytidine (ab142744) analog. Inhibits DNA methylation. Shows antitumor effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5503 Supplier Abcam Supplier No. AB14481550MG
MW 282.68 g/mol, Purity >99%. Nucleoside analog. 5-Azacytidine (ab142744) analog. Inhibits DNA methylation. Shows antitumor effects.
The product is subject to the following: Abcam Restricted Use Statement