Learn More
Selleck Chemical LLC NSC 319726 5mg 71555-25-4
SDP
Supplier:  Selleck Chemical LLC S71495MG
Encompass_Preferred
NSC319726 is a p53(R175) mutant reactivator, exhibits growth inhibition in cells expressing mutant p53, with IC50 of 8 nM for p53(R175) mutant, shows no inhibition for p53 wild-type cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4461