Learn More
Medchemexpress LLC NSC 357754 dihydrochloride | 98.0% | 10 MG
SDP
Catalog No. 5000295037
Encompass_Preferred
NSC 357754 dihydrochloride is a research-grade small molecule inhibitor of Claudin-2 and Claudin-15 that modulates epithelial barrier function by increasing transepithelial electrical resistance and reducing permeability to specific cations. It is provided in milligram pack sizes with reported high purity for laboratory studies of intestinal barrier physiology and related disease models.
  • Inhibits Claudin-2 and Claudin-15-mediated permeability.
  • Increases transepithelial electrical resistance in cell models.
  • Reduces permeability to specific cations.
  • Provided as a dihydrochloride salt in milligram pack sizes for research use.
  • High reported purity for reproducible experimental results.
  • Molecular weight 493.38 g/mol; molecular formula C26H22Cl2N4O2.
Catalog No. 50-002-95037 Supplier Medchemexpress LLC Supplier No. HY171152A10MG
NSC 357754 dihydrochloride is a research-grade small molecule inhibitor of Claudin-2 and Claudin-15 that modulates epithelial barrier function by increasing transepithelial electrical resistance and reducing permeability to specific cations. It is provided in milligram pack sizes with reported high purity for laboratory studies of intestinal barrier physiology and related disease models.
  • Inhibits Claudin-2 and Claudin-15-mediated permeability.
  • Increases transepithelial electrical resistance in cell models.
  • Reduces permeability to specific cations.
  • Provided as a dihydrochloride salt in milligram pack sizes for research use.
  • High reported purity for reproducible experimental results.
  • Molecular weight 493.38 g/mol; molecular formula C26H22Cl2N4O2.