Learn More
Medchemexpress LLC NSC 42834 (JAK2 Inhibitor V) | 195371-52-9 | C23H24N2O | 50 MG
SDP
Catalog No. 5000376268
Encompass_Preferred
NSC 42834 (JAK2 Inhibitor V) is a novel specific inhibitor of Jak2. It inhibits Jak2-V617F and Jak2-WT autophosphorylation in a dose-dependent manner without being cytotoxic to cells at concentrations that inhibit kinase activity. It selectively inhibits Jak2 kinase function without affecting Tyk2 or c-Src kinase function.
  • Inhibits Jak2-V617F and Jak2-WT autophosphorylation
  • Not cytotoxic to cells at kinase inhibiting concentrations
  • Selectively inhibits Jak2 kinase function
  • Inhibits proliferation of Jak2-V617F-expressing human erythroleukemia cell line
  • Reduces Jak2 and STAT3 tyrosine phosphorylation levels
  • Causes marked cell cycle arrest
Catalog No. 50-003-76268 Supplier Medchemexpress LLC Supplier No. HY1548050MG
NSC 42834 (JAK2 Inhibitor V) is a novel specific inhibitor of Jak2. It inhibits Jak2-V617F and Jak2-WT autophosphorylation in a dose-dependent manner without being cytotoxic to cells at concentrations that inhibit kinase activity. It selectively inhibits Jak2 kinase function without affecting Tyk2 or c-Src kinase function.
  • Inhibits Jak2-V617F and Jak2-WT autophosphorylation
  • Not cytotoxic to cells at kinase inhibiting concentrations
  • Selectively inhibits Jak2 kinase function
  • Inhibits proliferation of Jak2-V617F-expressing human erythroleukemia cell line
  • Reduces Jak2 and STAT3 tyrosine phosphorylation levels
  • Causes marked cell cycle arrest