Learn More
Abcam NSC-632839, ubiquitin isopeptidase inhibitor, 50MG
Catalog No. 502705585
Encompass_Preferred
MW 339.9 Da, Purity >98%. Nonselective ubiquitin isopeptidase inhibitor (IC₅₀ values are 45, 37 and 9.8 μM for USP2, USP7 and SENP2 respectively). Induces apoptosome-independent apoptosis and caspase activation. Antitumor agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5585 Supplier Abcam Supplier No. AB14459950MG
MW 339.9 Da, Purity >98%. Nonselective ubiquitin isopeptidase inhibitor (IC₅₀ values are 45, 37 and 9.8 μM for USP2, USP7 and SENP2 respectively). Induces apoptosome-independent apoptosis and caspase activation. Antitumor agent.
The product is subject to the following: Abcam Restricted Use Statement