Learn More
Medchemexpress LLC HY-100034 25mg Medchemexpress, NSC 663284 CAS:383907-43-5 Purity:>98%
SDP
Catalog No. 501931704
Encompass_Preferred
Medchemexpress, HY-100034 25mg NSC 663284 CAS:383907-43-5 NSC 663284 is a potent, cell-permeable, and irreversible Cdc25 dual specificity phosphatase inhibitor, has an IC 50 for Cdc25B2 of 0.21 μM. NSC 663284 exhibits mixed competitive kinetics against Cdc25A, Cdc25B(2), and Cdc25C with Ki values of 29, 95, and 89 nM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1704 Supplier Medchemexpress LLC Supplier No. HY10003425MG
Medchemexpress, HY-100034 25mg NSC 663284 CAS:383907-43-5 NSC 663284 is a potent, cell-permeable, and irreversible Cdc25 dual specificity phosphatase inhibitor, has an IC 50 for Cdc25B2 of 0.21 μM. NSC 663284 exhibits mixed competitive kinetics against Cdc25A, Cdc25B(2), and Cdc25C with Ki values of 29, 95, and 89 nM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.