Learn More
Abcam NSC 663284 (DA3003-1), Cdc25 inhibitor, 5MG
Catalog No. 502705785
Encompass_Preferred
MW 321.76 Da, Purity >98%. Potent, cell-permeable, irreversible non-selective Cdc25 inhibitor. Induces cell cycle arrest at both G1 and G2/M phase. Shows antiproliferative effects against tumor cell lines (IC₅₀ = 0.2-35 μM). Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5785 Supplier Abcam Supplier No. AB1444085MG
MW 321.76 Da, Purity >98%. Potent, cell-permeable, irreversible non-selective Cdc25 inhibitor. Induces cell cycle arrest at both G1 and G2/M phase. Shows antiproliferative effects against tumor cell lines (IC₅₀ = 0.2-35 μM). Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement