Learn More
Medchemexpress LLC HY-14967 5mg Medchemexpress, NSC 66811 CAS:6964-62-1 Purity:>98%
SDP
Catalog No. 502081669
Encompass_Preferred
Medchemexpress, HY-14967 5mg NSC 66811 CAS:6964-62-1 NSC 66811 is a MDM2-p53 inhibitor, with a Ki of 120 nM for binding to MDM2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1669 Supplier Medchemexpress LLC Supplier No. HY149675MG
Medchemexpress, HY-14967 5mg NSC 66811 CAS:6964-62-1 NSC 66811 is a MDM2-p53 inhibitor, with a Ki of 120 nM for binding to MDM2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.