Learn More
TARGETMOL CHEMICALS INC NSC 694623 5MG
SDP
Catalog No. 502579606
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NSC 694623 is a potent histone acetyltransferase (HAT) inhibitor with IC50 of 15.9 uM against recombinant HAT p300/CBP associated factor (PCAF). NSC 694623 shows anti-proliferative activity against some cancer cells which can be used for anti-cancer research. purity: 99%
Catalog No. 50-257-9606 Supplier TARGETMOL CHEMICALS INC Supplier No. T605585MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NSC 694623 is a potent histone acetyltransferase (HAT) inhibitor with IC50 of 15.9 uM against recombinant HAT p300/CBP associated factor (PCAF). NSC 694623 shows anti-proliferative activity against some cancer cells which can be used for anti-cancer research. purity: 99%