Learn More
Medchemexpress LLC HY-15146 100mg , NSC 74859 S3I-201 CAS:501919-59-1 Purity:98%
SDP
Catalog No. 501493550
Encompass_Preferred
Medchemexpress, HY-15146 100mg NSC 74859 S3I-201 CAS:501919-59-1 Formula:C16H15NO7S IC50: 86±33 μM (STAT3) Purity:98% NSC 74859 is a chemical probe inhibitor of Stat3 activity, selectively inhibits Stat3 DNA-binding activity in vitro with IC 50 of 86±33 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3550 Supplier Medchemexpress LLC Supplier No. HY15146100MG
Medchemexpress, HY-15146 100mg NSC 74859 S3I-201 CAS:501919-59-1 Formula:C16H15NO7S IC50: 86±33 μM (STAT3) Purity:98% NSC 74859 is a chemical probe inhibitor of Stat3 activity, selectively inhibits Stat3 DNA-binding activity in vitro with IC 50 of 86±33 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.