Learn More
Medchemexpress LLC NSC-87877 (disodium) | 56932-43-5 | 503.42 | 25 MG
SDP
Catalog No. 5000385552
Encompass_Preferred
NSC-87877 disodium is a potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases (SH-PTP2 and SH-PTP1). This compound also inhibits dual-specificity phosphatase 26 (DUSP26).
  • Potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases
  • Exhibits IC50 values of 0.318 μM for Shp2
  • Exhibits IC50 values of 0.355 μM for Shp1
  • Inhibits dual-specificity phosphatase 26 (DUSP26)
Catalog No. 50-003-85552 Supplier Medchemexpress LLC Supplier No. 25MGHY18756A
NSC-87877 disodium is a potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases (SH-PTP2 and SH-PTP1). This compound also inhibits dual-specificity phosphatase 26 (DUSP26).
  • Potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases
  • Exhibits IC50 values of 0.318 μM for Shp2
  • Exhibits IC50 values of 0.355 μM for Shp1
  • Inhibits dual-specificity phosphatase 26 (DUSP26)