Learn More
Medchemexpress LLC NSC-87877 (disodium) | 56932-43-5 | 98.0% | 503.42 | 50 MG
SDP
Catalog No. 5000385741
Encompass_Preferred
NSC-87877 disodium is a potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases, with IC50 values of 0.318 μM and 0.355 μM for Shp2 and Shp1, respectively. It also inhibits dual-specificity phosphatase 26 (DUSP26).
  • Potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases
  • Inhibits dual-specificity phosphatase 26 (DUSP26)
  • Inhibits DUSP26 function in NB cell lines
  • Results in increased p53 phosphorylation and activation
  • Possesses excellent anti-neuroblastoma activity in vivo
Catalog No. 50-003-85741 Supplier Medchemexpress LLC Supplier No. HY18756A50MG
NSC-87877 disodium is a potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases, with IC50 values of 0.318 μM and 0.355 μM for Shp2 and Shp1, respectively. It also inhibits dual-specificity phosphatase 26 (DUSP26).
  • Potent inhibitor of Shp2 and Shp1 protein tyrosine phosphatases
  • Inhibits dual-specificity phosphatase 26 (DUSP26)
  • Inhibits DUSP26 function in NB cell lines
  • Results in increased p53 phosphorylation and activation
  • Possesses excellent anti-neuroblastoma activity in vivo