Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NSC-87877 (disodium) | 56932-43-5 | 98.0% | 503.42 | C19H11N3Na2O7S2 | 10 MG
SDP

Catalog No. 5000294786
Encompass_Preferred

NSC-87877 disodium is the disodium salt form of a small-molecule inhibitor of protein tyrosine phosphatases SHP-1 and SHP-2, intended for research use in biochemical and cell-based studies. It is provided as a solid and characterized for biochemical potency and composition.

  • Potent inhibitor of SHP-1 and SHP-2 (IC50 ≈ 0.318 μM and 0.355 μM).
  • Provided as the disodium salt for improved solubility and handling.
  • High purity (98.0%) suitable for research applications.
  • Molecular weight 503.42 and formula C19H11N3Na2O7S2 for precise mass calculations.
  • Available in small milligram pack sizes for screening and assay development.

Catalog No. 50-002-94786 Supplier Medchemexpress LLC Supplier No. HY18756A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NSC-87877 disodium is the disodium salt form of a small-molecule inhibitor of protein tyrosine phosphatases SHP-1 and SHP-2, intended for research use in biochemical and cell-based studies. It is provided as a solid and characterized for biochemical potency and composition.

  • Potent inhibitor of SHP-1 and SHP-2 (IC50 ≈ 0.318 μM and 0.355 μM).
  • Provided as the disodium salt for improved solubility and handling.
  • High purity (98.0%) suitable for research applications.
  • Molecular weight 503.42 and formula C19H11N3Na2O7S2 for precise mass calculations.
  • Available in small milligram pack sizes for screening and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.