Learn More
Enzo Life Sciences NSC-95397 (5mg). CAS: 93718-83-3
SDP
Supplier:  Enzo Life Sciences BMLEI3090005
Encompass_Preferred
A potent and selective inhibitor of Cdc25 phosphatase (Ki's for Cdc25A, B and C are 32, 96 and 40 nM respectively). NSC-95397 inhibits the growth of several human tumor cell lines and blocks G2/M cell cycle transition. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (50mM) or ethanol (5mM). Long Term Storage: -20°C.
Catalog No. 50-201-0695