Learn More
Medchemexpress LLC HY-108543 5mg Medchemexpress, NSC 95397 CAS:93718-83-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085435MG
Encompass_Preferred
Medchemexpress, HY-108543 5mg NSC 95397 CAS:93718-83-3 NSC 95397 is a potent, selective Cdc25 dual specificity phosphatase inhibitor (Ki=32 nM (Cdc25A), 96 nM (Cdc25B), 40 nM (Cdc25C); IC50=22.3 nM (human Cdc25A), 56.9 nM (human Cdc25C), 125 nM (Cdc25B)). NSC 95397 inhibits mitogen-activated protein kinase phosphatase-1 (MKP-1) and suppresses proliferation and induces apoptosis in colon cancer cells through MKP-1 and ERK1/2 pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8574