Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSC 95397, Tocris Bioscience™

Catalog No. 154750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-475-0 50 mg
15-471-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-475-0 Supplier R&D Systems Supplier No. 1547/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Selective Cdc25 dual specificity phosphatase inhibitor

Potent and selective irreversible inhibitor of Cdc25 dual specificity phosphatases (Ki values are 32, 96 and 40 nM for inhibition of Cdc25A, -B and -C respectively). Displays 125 - 180-fold selectivity over VH1-related dual-specificity phosphatase and protein tyrosine phosphatase 1b. Inhibits carcinoma cell growth and blocks G2/M phase transition in vitro.

Chemical Identifiers

CAS 93718-83-3
Molecular Formula C14H14O4S2
Molecular Weight (g/mol) 310.382
InChI Key MAASHDQFQDDECQ-UHFFFAOYSA-N
Synonym 2,3-bis 2-hydroxyethylthio-1,4-naphthalenedione, 2,3-bis 2-hydroxyethylsulfanyl naphthalene-1,4-dione, 2,3-bis 2-hydroxyethyl sulfanyl naphthalene-1,4-dione, 1,4-naphthalenedione, 2,3-bis 2-hydroxyethyl thio, tocris-1547, lopac-n-1786, d0j1kh, 1, 2,3-bis 2-hydroxyethyl thio, nsc hplc, 2,3-bis 2-hydroxyethylthio naphthalene-1,4-dione
PubChem CID 262093
IUPAC Name 2,3-bis(2-hydroxyethylsulfanyl)naphthalene-1,4-dione
SMILES C1=CC=C2C(=C1)C(=O)C(=C(C2=O)SCCO)SCCO

Specifications

Quantity 50 mg
Formula Weight 310.38
Percent Purity >97%
Chemical Name or Material NSC 95397
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.