Learn More
Selleck Chemical LLC NSC12 5mg
SDP
Supplier:  Selleck Chemical LLC S79405MG
Encompass_Preferred
NSC12 (NSC 172285) is an orally available pan-FGF trap able to inhibit FGF2/FGFR interaction and endowed with promising antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3326