Learn More
Medchemexpress LLC HY-100791 5mg Medchemexpress, NSC23005 (sodium) CAS:1796596-46-7 Purity:>98%
SDP
Catalog No. 501931950
Encompass_Preferred
Medchemexpress, HY-100791 5mg NSC23005 (sodium) CAS:1796596-46-7 NSC23005 sodium is a novel and effective p18 inhibitor ( ED 50 =5.21 nM) in promoting Hematopoietic stem cells (HSCs) expansion in both murine and human models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1950 Supplier Medchemexpress LLC Supplier No. HY1007915MG
Medchemexpress, HY-100791 5mg NSC23005 (sodium) CAS:1796596-46-7 NSC23005 sodium is a novel and effective p18 inhibitor ( ED 50 =5.21 nM) in promoting Hematopoietic stem cells (HSCs) expansion in both murine and human models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.