Learn More
Selleck Chemical LLC NSC23005 Sodium
SDP
Catalog No. 5000002338
Encompass_Preferred
NSC 23005 is a small molecule inhibitor of p18 (ED50 5 21 nM)
Catalog No. 50-000-02338 Supplier Selleck Chemical LLC Supplier No. S65665MG
NSC 23005 is a small molecule inhibitor of p18 (ED50 5 21 nM)